- A Turning Point Towards Recovery: An Interview Study with Participants in the Culture and Health Programme for Clients with Long-Term Mental Health Disorders in Sweden.
A Turning Point Towards Recovery: An Interview Study with Participants in the Culture and Health Programme for Clients with Long-Term Mental Health Disorders in Sweden.
Issues in mental health nursing (2019-04-04)
Birgitta A Wästberg, Boel Sandström, Parvin Pooremamali
PMID30943065
ABSTRACT
The study aimed to explore participants' perceptions and experiences of the Culture and Health programme in Sweden for clients with long-term mental health disorders. A qualitative approach with interviews was applied. Grounded Theory guided the analysis and selection of informants. A total of 15 informants were interviewed. A core category 'A turning point in dealing with everyday life beyond the mental illness' with three categories: inner life, social life and occupational life emerged. A theory indicating the importance of asking clients about their expectations, was formulated. Further studies are warranted, including studies of effects.