Merck
  • Assessment of RNAlater® as a Potential Method to Preserve Bovine Muscle Proteins Compared with Dry Ice in a Proteomic Study.
Assessment of RNAlater® as a Potential Method to Preserve Bovine Muscle Proteins Compared with Dry Ice in a Proteomic Study.
Foods (Basel, Switzerland) (2019-02-16)
Yao Zhu, Anne Maria Mullen, Dilip K Rai, Alan L Kelly, David Sheehan, Jamie Cafferky, Ruth M Hamill
ABSTRACT
RNAlater® is regarded as a potential preservation method for proteins, while its effect on bovine muscle proteins has rarely been evaluated. Bovine muscle protein samples (n = 12) collected from three tender (Warner⁻Bratzler shear force: 30.02⁻31.74 N) and three tough (Warner⁻Bratzler shear force: 54.12⁻66.25 N) Longissimus thoracis et lumborum (LTL) samples, preserved using two different sampling preservation methods (RNAlater® and dry ice), at two post mortem time points (day 0 and day 14), were characterized using one-dimensional electrophoresis. Fourteen bands with molecular weights ranging from 15 to 250 kDa were verified, both in the dry ice and RNAlater® storage groups, at each time point, using image analysis. A shift from high to low molecular weight fragments, between day 0 and day 14, indicated proteolysis of the muscle proteins during post mortem storage. Liquid chromatography-tandem mass spectrometry (LC-MS/MS) analyses and database searching resulted in the identification of 10 proteins in four bands. Protein profiles of muscle preserved in RNAlater® were similar to those of muscle frozen on dry ice storage, both at day 0 and day 14. The results demonstrate that RNAlater® could be a simple and efficient way to preserve bovine muscle proteins for bovine muscle proteomic studies.