Merck
  • The cornified envelope of terminally differentiated human epidermal keratinocytes consists of cross-linked protein.
The cornified envelope of terminally differentiated human epidermal keratinocytes consists of cross-linked protein.
Cell (1977-06-01)
R H Rice, H Green
PMID890737
ABSTRACT
A small proportion of the protein of stratum corneum of human epidermal callus is insoluble even when boiled in solutions containing sodium dodecylsulfate and a reducing agent. This protein is present in the cornified envelope, a structure located beneath the plasma membrane. When cornified envelopes were dissolved by exhaustive proteolytic digestion and the products analyzed by chromatography, approximately 18% of the total lysine residues were found as the cross-linking dipeptide epsilon-(gamma-glutamyl) lysine. Labeled cornified envelope protein was synthesized by human epidermal keratinocytes allowed to differentiate terminally in culture. The extent of cross-linking, determined from the proportion of radioactive lysine in epsilon-(gamma-glutamyl) lysine after exhaustive proteolysis, was similar to that in stratum corneum. The properties of the cornified envelopes (insolubility in detergent and reducing agents, and solubility following proteolytic digestion) are readily explained by a structure consisting of a cross-linked protein lattice.