Leucovorin Calcium
Synonym(s):
Folinic acid calcium salt, Calcium folinate
Empirical Formula (Hill Notation):
C20H21CaN7O7
CAS Number:
Molecular Weight:
511.50
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3