Skip to Content
Merck
N-Nitroso-diethylamine European Pharmacopoeia (EP) Reference Standard

N-Nitrosodiethylamine (NDEA) solution

N-Nitroso-diethylamine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Diethylnitrosamine, N-Nitrosodiethylamine, N-Ethyl-N-nitrosoethanamine, N-Nitrosodiethylamine (NDEA) solution
Linear Formula:
(C2H5)2NNO
CAS Number:
Molecular Weight:
102.14

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique