Skip to Content
Merck
N-Isovalerylglycine ≥97.0% (GC)

N-Isovalerylglycine

N-Isovalerylglycine ≥97.0% (GC)
Synonym(s):
N-(3-Methyl-1-oxobutyl)glycine, N-(3-Methylbutanoyl)glycine, [(3-Methylbutanoyl)amino]acetic acid
Empirical Formula (Hill Notation):
C7H13NO3
CAS Number:
Molecular Weight:
159.18

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing