Skip to Content
ID
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Electrolyte solution
Synonym(s):
Lithium chloride solution
Linear Formula:
LiCl
CAS Number:
7447-41-8
Molecular Weight:
42.39
Products
Product Category
salts (1)
Brand
Supelco (2)
Formula Weight
Physical Form
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
56754
Electrolyte solution,
nonaqueous, 2 M LiCl in ethanol
Expand
View Pricing
62481
Electrolyte solution,
1 M in ethanol
Expand
View Pricing
16398
Electrolyte solution,
nonaqueous, LiCl in ethanol (saturated)
Expand
View Pricing