Skip to Content
Merck
BIOshell A160 Peptide C18 Column, 2 µm HPLC Column 2.0 μm particle size, L × I.D. 10 cm × 2.1 mm

BIOshell A160 Peptide C18 Column, 2 µm HPLC Column

BIOshell A160 Peptide C18 Column, 2 µm HPLC Column 2.0 μm particle size, L × I.D. 10 cm × 2.1 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique