Skip to Content
Merck
vibrant-m

Benzonase® Endonuclease ELISA Kit

vibrant-m
Synonym(s):
Endonuclease Detection Kit

Products

Product Category
Brand
Biological Source
Detection Method
Quality Segment
Application
Package Size
Technique