Skip to Content
Merck
Aquivion® E98-15S membrane sheet, stabilized CF3 polymer chain ends, PFSA eq. wt. 980 g/mole SO3H, L × W × thickness 18 cm × 18 cm × 150 μm

Aquivion® E98-15S

Aquivion® E98-15S membrane sheet, stabilized CF3 polymer chain ends, PFSA eq. wt. 980 g/mole SO3H, L × W × thickness 18 cm × 18 cm × 150 μm
Synonym(s):
Aquivion® SO3H
Linear Formula:
[CF2CF(OCF2CF2SO3H)]m[CF2CF2]n
CAS Number:

Products

Brand
Physical Form
Length (cm)