Skip to Content
Merck
Aflatoxin B1 from Aspergillus flavus from Aspergillus flavus

Aflatoxin B1 from Aspergillus flavus

Aflatoxin B1 from Aspergillus flavus from Aspergillus flavus
Empirical Formula (Hill Notation):
C17H12O6
CAS Number:
Molecular Weight:
312.27
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Feature
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing