Skip to Content
Merck
Deferoxamine mesylate salt ≥92.5% (TLC), powder

Deferoxamine mesylate salt

Deferoxamine mesylate salt ≥92.5% (TLC), powder
Synonym(s):
DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt
Empirical Formula (Hill Notation):
C25H48N6O8 · CH4O3S
CAS Number:
Molecular Weight:
656.79
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing