Imatinib mesylate
Synonym(s):
4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-phenyl]benzamide methanesulfonate, 4-[(4-Methylpiperazin-1-yl)methyl]-N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)benzamide mesylate, CGP 57148, Genfatinib, Glivec, Imatinib methanesulfonate, STI-571
Empirical Formula (Hill Notation):
C29H31N7O · CH4O3S
CAS Number:
Molecular Weight:
589.71
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing