Sodium borohydride
Synonym(s):
Sodium tetrahydridoborate, Sodium borohydride
Linear Formula:
NaBH4
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
Showing 1-11 of 11
Compare
Product Number
Description
Pricing