Skip to Content
Merck
L-Glutathione reduced BioXtra, ≥98.0%

L-Glutathione reduced

L-Glutathione reduced BioXtra, ≥98.0%
Synonym(s):
γ-L-Glutamyl-L-cysteinyl-glycine, GSH, Glutathione (reduced), Glutathione, Reduced, Free Acid
Linear Formula:
H2NCH(CO2H)CH2CH2CONHCH(CH2SH)CONHCH2CO2H
CAS Number:
Molecular Weight:
307.32
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Optical Isomer
Technique