Skip to Content
Merck
1L Tedlar® Gas Sampling Bag for EPA TCLP Method pkg of 10 bags

1L Tedlar® Gas Sampling Bag for EPA TCLP Method

1L Tedlar® Gas Sampling Bag for EPA TCLP Method pkg of 10 bags
Synonym(s):
Tedlar® Gas Sampling Bag

Products

Product Category
Agency
Length (in)
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing