![1,8-Diazabicyclo[5.4.0]undec-7-ene solution 1 M in THF](/deepweb/assets/sigmaaldrich/product/structures/120/564/5b373e23-1624-489c-8efb-692de0f96ffb/640/5b373e23-1624-489c-8efb-692de0f96ffb.png)
1,8-Diazabicyclo[5.4.0]undec-7-ene solution
![1,8-Diazabicyclo[5.4.0]undec-7-ene solution 1 M in THF](/deepweb/assets/sigmaaldrich/product/structures/120/564/5b373e23-1624-489c-8efb-692de0f96ffb/640/5b373e23-1624-489c-8efb-692de0f96ffb.png)
Synonym(s):
DBU solution
Empirical Formula (Hill Notation):
C9H16N2
Molecular Weight:
152.24
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing