Sodium phosphate monobasic dihydrate
Synonym(s):
Sodium dihydrogen phosphate dihydrate, Sodium phosphate monobasic dihydrate
Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Showing 1-7 of 7
Compare
Product Number
Description
Pricing