Lysozyme from chicken egg white
Synonym(s):
Mucopeptide N-acetylmuramoylhydrolase, Muramidase, Lysozyme from chicken egg white, Lysozyme, Chicken Egg White
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Manufacturer
Physical Form
Recombinant Host
Tissue
Chemical Composition
Quality Segment
Application
Reference Material Type
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing