Skip to Content
Merck
All Photos(1)

Key Documents

ABE70-I

Sigma-Aldrich

Anti-BAF180/PBRM1

Synonym(s):

Protein polybromo-1;hPB1;BRG1-associated factor 180;Polybromo-1D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

mol wt

calculated mol wt 192.95 kDa
observed mol wt ~200 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PBRM1(55193)

General description

Protein polybromo-1 (UniProt: Q86U86; also known as hPB1, BRG1-associated factor 180, BAF180, Polybromo-1D) is encoded by the PBRM1 (also known as BAF180, PB1) gene (Gene ID: 55193) in human. BAF180 is a widely expressed, bromodomain containing nuclear protein that is an essential component of the SWI/SNF-B (PBAF) chromatin remodeling complex. It is involved in transcriptional activation and repression of select genes by chromatin remodeling. It contains six bromodomains, two bromo-adjacent homology (BAH) domains (aa 956-1074 and 1156-1272), and one HMG-box (aa 1379-1447). The bromodomains recognize and bind to acetylated lysines and BAH domains mediate protein-protein interaction. Its HMG-box domain binds to DNA. BAF180 acts as a negative regulator of cell proliferation. It plays a role in DNA repair mechanisms and acts as a tumor suppressor. It is also involved in silencing transcription near double-stranded DNA breaks (DSB). The BAH domains of BAF180 are shown to be essential for ubiquitination of PCNA. BAF180 depleted cells can not produce proper levels of ubiquitinated PCNA that leads to the inability to replicate past DNA lesions. Mutations in PBRM1 gene have been linked to high incidence of renal cell carcinoma. Nine isoforms of BAF180 have been described that are produced by alternative splicing. (Ref.: Hopson, S., and Thompson, MJ. (2017). ACS Chem. Biol. 12(10); 2482-2490; Varela, I., et al. (2011). Nature. 469(7331); 539-542).

Specificity

This rabbit polyclonal antibody specifically detects Protein polybromo-1 (BAF180). It targets an epitope within the C-terminal half.

Immunogen

GST-tagged recombinant fragment corresponding to 740 amino acids from the C-terminal half of human BAF180

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Protein polybromo-1 (BAF180) in Jurkat cell lysate.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service