Merck
HomePeptide SynthesisMerrifield Peptide Resins
Merrifield Peptide Resins
Rarely does research that is ultimately honored by a Nobel Prize result in specific products of particular use to the scientific community, as have these peptide resins, available from us.
These chloromethylated styrene/divinyl benzene copolymers have been crucial to the development of techniques and technology leading to the routine, convenient synthesis of peptides.