IC Separation of Chloride, Fluoride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 5
![IC Separation of Chloride, Fluoride, Nitrite, Bromide, Nitrate, Phosphate, and Sulfate Ions on Metrohm Metrosep A Supp 5 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/265/089/125685f3-f985-4aff-ac1c-94f5882593d5/800/125685f3-f985-4aff-ac1c-94f5882593d5.jpg)
Materials
mobile phase component
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
Metrosep A Supp 5, 15 cm x 4 mm I.D., 5 μm particles (Metrohm 6.1006.520)
mobile phase
[A] 3.2 mM sodium carbonate; [B] 1.0 mM sodium bicarbonate
flow rate
0.7 mL/min
column temp.
25 °C
detector
conductivity
injection
20 μL
sample
chloride, fluoride (2 mg/L), nitrite (5 mg/L), bromide, nitrate, phosphate, sulfate (10 mg/L)