Merck
  • The role of the endocannabinoid system in the antihyperalgesic effect of Cedrus atlantica essential oil inhalation in a mouse model of postoperative pain.
The role of the endocannabinoid system in the antihyperalgesic effect of Cedrus atlantica essential oil inhalation in a mouse model of postoperative pain.
Journal of ethnopharmacology (2017-09-18)
Aline Armiliato Emer, Nathalia Nahas Donatello, Ana Paula Batisti, Luiz Augusto Oliveira Belmonte, Adair R S Santos, Daniel F Martins
ABSTRACT
Cedar is part of the phylum of conifers, and it's essential oil has been used for therapeutic purposes since ancient times. In our previous study, we have demonstrated that the inhalation of the Cedrus atlantica essential oil (CaEO) induces an antihyperalgesic effect in a model of postoperative pain. But the mechanism that underlies this effect is not yet fully known. This study investigates the involvement of the endocannabinoid system in the antihyperalgesic effect produced by the inhalation of CaEO in a post operative pain model. Male Swiss mice (25-35±2g) were subjected to plantar incision. To assess the involvement of the endocannabinoid system, two different approaches were made: (1) by administering antagonists to the CB The antihyperalgesic effect of CaEO inhalation was prevented by pretreatment with AM281 or AM630 given by i.p. and i.t., but not i.pl. Additionally, in mice pretreated with FAAH or the MAGL inhibitors, the antihyperalgesic effect of CaEO inhalation was significantly longer, which demonstrates the involvement of the endocannabinoid system in the antihyperalgesic effect of CaEO inhalation in a preclinical model of postoperative pain. The present study shows that CaEO inhalation exerts an antihyperalgesic effect, possibly by the activation of the endocannabinoid system in a preclinical model of postoperative pain. It could be a new alternative to treat pain in a clinical environment.
Sigma-Aldrich
AM281, ≥98% (HPLC)
Sigma-Aldrich
AM630, ≥90% (HPLC)