Merck
Pharmacological effects of cannabinoids on learning and memory in
The Journal of experimental biology (2017-09-01)
Hiroshi Sunada, Takayuki Watanabe, Dai Hatakeyama, Sangmin Lee, Jeremy Forest, Manabu Sakakibara, Etsuro Ito, Ken Lukowiak
ABSTRACT
Cannabinoids are hypothesized to play an important role in modulating learning and memory formation. Here, we identified mRNAs expressed in
Sigma-Aldrich
AM251, >98% (HPLC), solid