Merck
  • MYCN is an immunosuppressive oncogene dampening the expression of ligands for NK-cell-activating receptors in human high-risk neuroblastoma.
MYCN is an immunosuppressive oncogene dampening the expression of ligands for NK-cell-activating receptors in human high-risk neuroblastoma.
Oncoimmunology (2017-07-07)
Elisa Brandetti, Irene Veneziani, Ombretta Melaiu, Annalisa Pezzolo, Aurora Castellano, Renata Boldrini, Elisa Ferretti, Doriana Fruci, Lorenzo Moretta, Vito Pistoia, Franco Locatelli, Loredana Cifaldi
ABSTRACT
Neuroblastoma (NB) is the most common extracranial solid tumor occurring in childhood. Amplification of the