Merck
  • Generation of a felinized swine endothelial cell line by expression of feline decay-accelerating factor.
Generation of a felinized swine endothelial cell line by expression of feline decay-accelerating factor.
PloS one (2015-02-12)
Luna Izuhara, Norifumi Tatsumi, Shuji Miyagawa, Satomi Iwai, Masahito Watanabe, Shuichiro Yamanaka, Yuichi Katsuoka, Hiroshi Nagashima, Hirotaka J Okano, Takashi Yokoo
ABSTRACT
Embryonic stem cell research has facilitated the generation of many cell types for the production of tissues and organs for both humans and companion animals. Because ≥30% of pet cats suffer from chronic kidney disease (CKD), xenotransplantation between pigs and cats has been studied. For a successful pig to cat xenotransplant, the immune reaction must be overcome, especially hyperacute rejection. In this study, we isolated the gene for feline decay-accelerating factor (fDAF), an inhibitor of complement proteins, and transfected a swine endothelial cell line with fDAF to "felinize" the pig cells. These fDAF-expressing cells were resistant to feline serum containing anti-pig antibodies, suggesting that felinized pig cells were resistant to hyperacute rejection. Our results suggest that a "felinized" pig kidney can be generated for the treatment of CKD in cats in the future.