Merck
  • Frequency and Effects of Excess Dosing of Anticoagulants in Patients ≤55 Years With Acute Myocardial Infarction Who Underwent Percutaneous Coronary Intervention (from the VIRGO Study).
Frequency and Effects of Excess Dosing of Anticoagulants in Patients ≤55 Years With Acute Myocardial Infarction Who Underwent Percutaneous Coronary Intervention (from the VIRGO Study).
The American journal of cardiology (2015-05-06)
Aakriti Gupta, Philip Chui, Shengfan Zhou, John A Spertus, Mary Geda, Nancy Lorenze, Ike Lee, Gail D' Onofrio, Judith H Lichtman, Karen P Alexander, Harlan M Krumholz, Jeptha P Curtis
ABSTRACT
Excess dosing of anticoagulant agents has been linked to increased risk of bleeding after percutaneous coronary intervention (PCI) for women compared with men, but these studies have largely included older patients. We sought to determine the prevalence and gender-based differences of excess dosing of anticoagulants including glycoprotein IIb/IIIa inhibitors, bivalirudin, and unfractionated heparin in young patients with acute myocardial infarction who underwent PCI and to examine its association with bleeding. Of 2,076 patients enrolled in the Variation in Recovery: Role of Gender on Outcomes of Young Acute Myocardial Infarction Patients study who underwent PCI, we abstracted doses of unfractionated heparin, bivalirudin, and glycoprotein IIb/IIIa inhibitors administered during PCI from the medical records. At least 47.2% received at least 1 excess dose of an anticoagulant, which did not differ by gender. We used logistic regression to determine the predictors of excess dosing and the association of excess dosing with bleeding. In multivariable analysis, only lower body weight and younger age were significant predictors of excess dosing. Bleeding was higher in young women who received excess dosing versus those who did not (9.3% vs 6.0%, p = 0.03) but was comparable among men (5.2% vs 5.9%, p = 0.69) in univariate analysis. In multivariable analysis, there was a trend to an association between excess dosing and bleeding (odds ratio 1.33, 95% confidence interval 0.92 to 1.91) although not statistically significant. In conclusion, approximately half of the patients received excess dosing of anticoagulant drugs during PCI, which did not vary based on gender. There was a trend toward an association between excess dosing and increased bleeding, although not statistically significant.
Sigma-Aldrich
Bivalirudin trifluoroacetate salt, ≥97% (HPLC)