Merck
  • Microvascular protective role of pericytes in melatonin-treated spinal cord injury in the C57BL/6 mice.
Microvascular protective role of pericytes in melatonin-treated spinal cord injury in the C57BL/6 mice.
Chinese medical journal (2014-08-26)
Yingli Jing, Qingbin Wu, Xiaochen Yuan, Bingwei Li, Mingming Liu, Xiaoyan Zhang, Shuying Liu, Hongwei Li, Ruijuan Xiu
ABSTRACT
Pericytes, located on microvessels, help to maintain vascular stability and blood-brain barrier integrity. The influence of pericytes on microvessels after spinal cord injury (SCI) is less clear. Therefore, the aim of this study was to investigate whether pericytes took a protective effect on microvessels in melatonin-treated SCI. C57BL/6 mice were randomly divided into three groups: sham group, SCI group, and melatonin group (n = 27 per group). Functional recovery was evaluated using the Basso Mouse Scale. Motor neurons were observed using hematoxylin and eosin staining. Pericyte coverage was analyzed using immunofluorescence. Permeability of blood-spinal cord barrier (BSCB) was assessed by administration of Evan's Blue. Protein levels of occludin, aquaporin-4 (AQP4), angiopoietin-1 (Ang1), intercellular cell adhesion molecule-1 (ICAM-1), Bcl-2, and Bax were determined using Western blotting. Mimicking the pathological conditions of SCI, melatonin-treated primary pericytes were subjected to oxygen-glucose deprivation/reperfusion (OGD/R). Secretion of Ang1 was analyzed using an enzyme-linked immunosorbent assay, and the expression of ICAM-1 was detected by immunofluorescence. Melatonin treatment improved locomotor functional outcome and rescued motor neurons. Pericyte coverage was significantly reduced after SCI; melatonin treatment alleviated the loss of pericyte coverage and rescued perfused microvessels 7 days after injury. The permeability of BSCB and loss of occludin were attenuated, and edema formation and upregulation of AQP4 were inhibited, after melatonin treatment. The expression of Ang1 and Bcl-2 was improved, while the expression of ICAM-1 and Bax was inhibited, in melatonin-treated SCI mice. Furthermore, the secretion of Ang1 was increased and the expression of ICAM-1 was inhibited in melatonin-treated pericytes after OGD/R. Melatonin ameliorated the loss of blood vessels and disruption of BSCB to exert a protective effect on SCI, which might be mediated by increased pericyte coverage. The upregulation of Ang1 in pericytes could inhibit inflammation and apoptosis to protect the microvessels.