Merck
  • Hrs is associated with STAM, a signal-transducing adaptor molecule. Its suppressive effect on cytokine-induced cell growth.
Hrs is associated with STAM, a signal-transducing adaptor molecule. Its suppressive effect on cytokine-induced cell growth.
The Journal of biological chemistry (1998-01-31)
H Asao, Y Sasaki, T Arita, N Tanaka, K Endo, H Kasai, T Takeshita, Y Endo, T Fujita, K Sugamura
ABSTRACT
We previously reported a new type of signal-transducing adaptor molecule, STAM, which was shown to be involved in cytokine-mediated intracellular signal transduction. In this study, we molecularly cloned a 110-kDa phosphotyrosine protein inducible by stimulation with interleukin 2 (IL-2). The 110-kDa molecule was found to be a human counterpart of mouse Hrs (hepatocyte growth factor-regulated tyrosine kinase substrate) and to be associated with STAM. Tyrosine phosphorylation of Hrs is induced rapidly after stimulation with IL-2 and granulocyte-macrophage colony-stimulating factor as well as hepatocyte growth factor. The mutual association sites of Hrs and STAM include highly conserved coiled-coil sequences, suggesting that their association is mediated by the coiled-coil structures. Exogenous introduction of the wild-type Hrs significantly suppressed DNA synthesis upon stimulation with IL-2 and granulocyte-macrophage colony-stimulating factor, while the Hrs mutant deleted of the STAM-binding site lost such suppressive ability. These results suggest that Hrs counteracts the STAM function which is critical for cell growth signaling mediated by the cytokines.