Merck
Restoring a metabolic pathway.
ACS chemical biology (2008-10-22)
John P Richard
ABSTRACT
Gluconeogenesis is blocked in a strain of Escherichia coli that is deficient in triosephosphate isomerase, but it was restored by the insertion of a plasmid coding for an L-glyceraldehyde 3-phosphate reductase (YghZ). This reductase provides a "bypass" that produces dihydroxyacetone phosphate (DHAP) by the consecutive enzyme-catalyzed reduction of L-glyceraldehyde 3-phosphate ( L-GAP) by NADPH to give L-glycerol 3-phosphate and reoxidation by NAD(+) catalyzed by endogenous L-glycerol 3-phosphate dehydrogenase to give DHAP. The origin of cellular L-GAP remains to be determined.
Sigma-Aldrich
Dihydroxyacetone phosphate lithium salt, ≥95.0% (TLC)