Merck
  • Chemiluminescence characteristics of cumarin derivatives as blue fluorescers in peroxyoxalate-hydrogen peroxide system.
Chemiluminescence characteristics of cumarin derivatives as blue fluorescers in peroxyoxalate-hydrogen peroxide system.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-03-28)
Mohammad Javad Chaichi, Ali Reza Karami, Abbas Shockravi, Mojtaba Shamsipur
ABSTRACT
The chemiluminescence characteristics of seven different cumarin derivatives were studied in detail. The fluorescence and chemiluminescence spectra were compared; all cumarins used were found to act as blue fluorescers. The intensity and kinetic parameters for the chemiluminescent systems were evaluated from computer fitting of the resulting intensity-time plots. Among different cumarin derivatives used, 7-amino-4-trifluoromethylcumarin revealed the most promising characteristics as an efficient blue fluorescent emitter.
Sigma-Aldrich
7-Amino-4-(trifluoromethyl)coumarin, ≥99%
Pricing and availability is not currently available.