Merck
  • Platelet adhesion and complement activation studies on poly(N-alkyl mono and disubstituted) acrylamide derivatives.
Platelet adhesion and complement activation studies on poly(N-alkyl mono and disubstituted) acrylamide derivatives.
Biomaterials (1999-02-27)
R Bahulekar, N Tamura, S Ito, M Kodama
ABSTRACT
Poly(N-alkyl mono and disubstituted) acrylamide derivatives were synthesized from poly(acryloyl chloride) by monomer analogous reaction. The polymers were characterized by FTIR-ATR and GPC. The contact angle measurements were performed to evaluate hydrophobic/hydrophilic characters of these polymers. The N-alkyl substituents changed contact angle between 55 and 75 degrees. In vitro platelet adhesion studies showed that surfaces of poly(N-alkyl substituted) acrylamides are prone to adhere platelets. Platelet spreading was more on poly(N-benzyl-N-ethyl acrylamide) surfaces in comparison to that on poly(N-benzyl-N-propionic acid ethyl ester acrylamide) and poly(N-benzyl acrylamide) surfaces. As a result of modification of amino group with N-alkyl substituents, the activations of C3a and C5a complements were suppressed 9-20% and 5-6% of native poly(acrylamides), respectively.
Sigma-Aldrich
Acryloyl chloride, ≥97%, contains ~400 ppm phenothiazine as stabilizer
Sigma-Aldrich
Acryloyl chloride, 97.0%, contains <210 ppm MEHQ as stabilizer