Merck
  • Inhibition of breast cancer cell proliferation with anti-microRNA oligonucleotides flanked by interstrand cross-linked duplexes.
Inhibition of breast cancer cell proliferation with anti-microRNA oligonucleotides flanked by interstrand cross-linked duplexes.
Nucleosides, nucleotides & nucleic acids (2019-10-05)
Sho Okumura, Yu Hirano, Yasuo Komatsu
ABSTRACT
Breast cancer is the most frequent cancer affecting women worldwide. Traditional chemotherapy, hormone therapy, and targeted therapy are used for breast cancer treatment. However, breast cancer is a heterogeneous disease, and patients often develop drug resistance. Therefore, various new therapeutic strategies have been investigated, including microRNA regulation. Anti-microRNA oligonucleotides (AMOs) are one of the most potent agents in oligonucleotide therapy. The inhibition activity of an AMO can be increased by flanking its single-stranded antisense sequence (the widely used structure for AMOs) with interstrand cross-linked duplexes (CLDs). An extrastable CLD improves nuclease resistance and stabilizes hybridization with a target. This study investigated the effects of anti-microRNA-21 (miR-21) AMO modified with CLDs on breast cancer cells without using reporter assay. The CLD-modified AMO suppressed breast cancer cell proliferation for a long duration compared to other types of AMOs. In addition, it expectedly up-regulated the miR-21-controlled expression of tumor suppressor genes. Therefore, an AMO flanked by CLDs can be a promising strategy for breast cancer treatment.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder