- The study of ameliorative effect of dietary supplementation of vitamin C, vitamin E, and tryptophan on Labeo rohita (Cyprinidae) fry exposed to intense light.
The study of ameliorative effect of dietary supplementation of vitamin C, vitamin E, and tryptophan on Labeo rohita (Cyprinidae) fry exposed to intense light.
The stress ameliorating effect of dietary supplementation of vitamin C, vitamin E, and tryptophan on rohu Labeo rohita fry was evaluated. Rohu fry (1.1 ± 0.03 g) were cultured under five different feeding regimes: enriched with 0.08% vitamin C (D1), 0.02% vitamin E (D2), 1.42% tryptophan (D3), a combination of these three ingredients at similar doses (D4), and control diet (D5). Rohu fry of D5 were divided into two groups-exposed to experimental light (D5FL) and ambient light (114 ± 4 lx, D5AL). All fry (except D5AL) were exposed at light intensity of 3442 ± 648 lx. Feeding of rohu with enriched diets significantly (P < 0.05) enhanced the survival rate and average weight. A 15-25% higher survival and 1.3-1.8-fold higher average weight were recorded in rohu fed with enriched diet compared to D5FL treatment. Supplementation of vitamin C in diet (D1) of rohu resulted in 4.1-fold and 6.9-fold higher nitric oxide synthase and reduced glutathione (GSH) levels, respectively compared to the D5FL treatment. The tryptophan-enriched diet (D3) showed 5.8-fold higher melatonin and 4.4-fold lower cortisol levels in rohu compared to the D5FL treatment. Significantly (P < 0.05) higher nitric oxide synthase, GSH and melatonin, and lower cortisol, glucose, thiobarbituric acid reactive substances, carbonyl protein, glutathione S-transferase, and glutathione peroxidase levels were found in D4 diet fed rohu compared to the other treatments. Reduced level of stress in D4 treatment resulted in best performance of rohu in terms of less swimming activity and higher survival and growth compared to the other treatments.