Merck
  • NIX-mediated mitophagy protects against proteinuria-induced tubular cell apoptosis and renal injury.
NIX-mediated mitophagy protects against proteinuria-induced tubular cell apoptosis and renal injury.
American journal of physiology. Renal physiology (2018-09-13)
Dan Xu, Panpan Chen, Bao Wang, Yanzhe Wang, Naijun Miao, Fan Yin, Qian Cheng, Zhuanli Zhou, Hongyan Xie, Li Zhou, Jun Liu, Xiaoxia Wang, Roy Zent, Limin Lu, Wei Zhang
ABSTRACT
Proteinuria, the most common symptom of renal injury, is an independent factor for renal tubular injury. However, the underlying mechanism remains to be fully elucidated. Mitochondrion is an important target for proteinuria-induced renal tubular cell injury. Insufficient mitophagy exacerbates cell injury by initiating mitochondrial dysfunction-related cell apoptosis. In the experiment, the role of NIP3-like protein X (NIX)-mediated mitophagy was investigated in proteinuria-induced renal injury. In this study, we demonstrated that NIX expression was reduced in renal tubules and correlated with the decline of estimated glomerular filtration rate and increase of the proteinuria in patients. In proteinuric mice, NIX-mediated mitophagy was significantly suppressed. Meanwhile, the proteinuric mice exhibited renal dysfunction, increased mitochondrial fragmentation, and tubular cell apoptosis. Overexpression of NIX attenuated those disruptions in proteinuric mice. In cultured renal tubular epithelial cells, albumin induced a decrease in NIX-mediated mitophagy and an increase in cell apoptosis. Overexpression of NIX attenuated albumin-induced cell apoptosis, whereas NIX siRNA aggravated these perturbations. These results indicate that proteinuria suppresses NIX-mediated mitophagy in the renal tubular epithelial cell, which triggers the cell undergoing mitochondria-dependent cell apoptosis. Collectively, our finding suggests that restoration of NIX-mediated mitophagy might be a novel therapeutic target for alleviating proteinuria-induced kidney injury.