Glycine
Synonym(s):
Aminoacetic acid, Aminoethanoic acid, Glycocoll
Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
pK Value
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing