Skip to Content
Merck
Colchicine British Pharmacopoeia (BP) Reference Standard

Colchicine

Colchicine British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Colchicine, (S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide
Empirical Formula (Hill Notation):
C22H25NO6
CAS Number:
Molecular Weight:
399.44
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique