(−)-Borneol
Synonym(s):
endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol, (−)-Borneol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3