Skip to Content
Merck
Mesalazine impurity A European Pharmacopoeia (EP) Reference Standard

4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K)

Mesalazine impurity A European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-Aminophenol, 4-Hydroxyaniline
Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique