StableCell™ DMEM/F12
Synonym(s):
Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, DME/F-12, 1:1 mixture
Products
Product Category
Brand
Physical Form
Glucose (g/L)
HEPES (mM)
Sodium Bicarbonate
Phenol Red
Sodium Pyruvate (g/L)
Stable Glutamine
Quality Segment
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing