Skip to Content
Merck
Sodium hydroxide solution min. 27% (1.30) for analysis (for the determination of nitrogen) EMSURE®

Sodium hydroxide solution

Sodium hydroxide solution min. 27% (1.30) for analysis (for the determination of nitrogen) EMSURE®
Synonym(s):
Sodium hydroxide solution
Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Package Size
Reaction Type
Technique
Showing 1-20 of 37
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2