Skip to Content
Merck
Plasminogen activator inhibitor 1 (PAI-1) human recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Plasminogen activator inhibitor 1 (PAI-1) human

Plasminogen activator inhibitor 1 (PAI-1) human recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonym(s):
PAIE, PLANH1, Plasminogen activator inhibitor, β-migrating endothelial cell-derived type, Serpin E1 PAI-1, Serpine 1
CAS Number:

Products

Product Category
Brand
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Showing 1-1 of 1