N-Methyl-4-nitrophenethylamine hydrochloride
Synonym(s):
N-Methyl-2-(4-nitrophenyl)ethylamine hydrochloride
Empirical Formula (Hill Notation):
C9H12N2O2 · HCl
CAS Number:
Molecular Weight:
216.66
Products
Product Category
Brand
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing