Skip to Content
HU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thrombin from bovine plasma
Synonym(s):
Factor IIa, Thrombin, Bovine, Thrombin, Bovine, High Activity, Thrombin, Thrombin, Plasminogen-Free, Bovine
CAS Number:
9002-04-4
EC Number:
232-648-7
Products
Brand
Sigma-Aldrich (8)
Biological Source
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
T4648
Thrombin from bovine plasma,
lyophilized powder, 40-300 NIH units/mg protein (biuret)
Expand
View Pricing
T6634
Thrombin from bovine plasma,
lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
Expand
View Pricing
T7326
Thrombin from bovine plasma,
≥60 NIH units/mg protein (biuret)
Expand
View Pricing
605157
Thrombin from bovine plasma,
Expand
View Pricing
604980
Thrombin from bovine plasma,
Expand
View Pricing
1.12374
Thrombin from bovine plasma,
from bovine plasma, 50 NIH units/mg dry wt
Expand
View Pricing
605160
Thrombin from bovine plasma,
Expand
View Pricing
SRE0003
Thrombin from bovine plasma,
Suitable for manufacturing of diagnostic kits and reagents
Expand
View Pricing