Skip to Content
HU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Charcoal activated
Synonym(s):
Charcoal activated
Linear Formula:
C
CAS Number:
7440-44-0
Molecular Weight:
12.01
Products
Brand
SAFC (1)
Sigma-Aldrich (2)
Supelco (6)
Formula Weight
Physical Form
Agency
Quality Segment
Application
Particle Size (µm)
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
1.02184
Charcoal activated,
powder extra pure
Expand
View Pricing
1.02186
Charcoal activated,
for analysis
Expand
View Pricing
1.02183
Charcoal activated,
pure
Expand
View Pricing
901931
Charcoal activated,
meets analytical specifications of USP
Expand
View Pricing
1.02514
Charcoal activated,
granular about 1.5 mm extra pure
Expand
View Pricing
901934
Charcoal activated,
Expand
View Pricing
1.02555
Charcoal activated,
Ph Eur
Expand
View Pricing
1.02005
Charcoal activated,
for the determination of AOX (shaking method)
Expand
View Pricing
1.02216
Charcoal activated,
for the determination of AOX
Expand
View Pricing