Skip to Content
Merck
vibrant-m

L-α-Phosphatidylethanolamine from egg yolk

vibrant-m
Synonym(s):
1,2-Diacyl-sn-glycero-3-phosphoethanolamine, L-α-Cephalin
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Optical Isomer