Skip to Content
Merck
Nicotinamide ≥98.5% (HPLC)

Nicotinamide

Nicotinamide ≥98.5% (HPLC)
Synonym(s):
Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP, Nicotinamide
Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Special Grade
Technique