Z220248
1 of 10
Sigma-Aldrich
Z166898
Aldrich® 气敏化学玻璃实验室套件
Z150789
Glass tubing cutter replacement cutting wheels
Z220418
Cold-trap for isothermic evaporation
Z167444
Vacuum distillation adapter with threaded 14/20 joints
Z555525
Aldrich® AtmosBag手套袋
Z515981
Schlenk reaction and storage tube
Z220302
Reaction flask with glass stopcock on side-arm
SYNF351430M
Synthware™ fritted filter tube with glass stopcocks
Z106925
Sealing tape for AtmosBag
Z174238
Aldrich® two-piece vacuum trap
G
female joint: ST/NS14/20male joint: ST/NS14/20
玻璃耗材
批号
在文件库中查找您最近购买产品的文档。
访问文档库
1 of 7
Z173134
Aldrich ® Schlenk 型适配器
Z768278
Aldrich® high-vacuum glass stockcock, T-bore
Z549479
Stopcock Adapter
Z101370
Aldrich® septum-inlet adapter with stopcock
Z230855
Aldrich® Schlenk-type male joint cap
Z173061
Solvent storage flask with glass stopcock on side-arm
Z173231
Reaction and storage flask, Kjeldahl, with glass stopcock on side-arm