HomeGelatin Coating Protocol
Gelatin Coating Protocol
Important points to remember before the start
- Ensure the cells are healthy and in an adequate number.
- Gelatin solutions need to completely liquefy at 37 °C before coating.
- Gelatin solutions may be stored at 4 - 8 °C indefinitely; elevated temperatures cause hydrolysis and loss of integrity.
- The typical surface coverage concentration is 100 – 200 ug/cm2. Surface coverage concentration may differ with the cells being cultured. Optimal conditions for attachment must be determined for each cell line and application.
- Do not over-dry post coating as this disrupts the structure of gelatin and affects the attachment of cells.
Gelatin coating protocol for culture ware
- Prepare a 2% (w/v) solution by dissolving gelatin in tissue culture grade water.
- Sterilize by autoclaving at 121 °C, 15 psi for 30 minutes.
- Coat culture surface with 5-10 µL gelatin solution/cm2 (i.e., 100-200 µg/cm2).
- Allow to dry at least 2 hours before introducing cells and medium.
- Alternatively, a gelatin solution can be used to directly coat the culture ware.
Table 1: Gelatins for cell attachment
Click to learn more about Gelatins
Explore our 3D cell culture tools
Troubleshoot your cell culture challenges
Click for consistency in cell culture
Frequently Asked Questions (FAQs)
- What is the purity of gelatin products?
We do not determine the purity of fibronectin solutions. All our fibronectin powders are 70 - 90% pure.
- What is the best solvent to dilute gelatin powders?
The solvents that are compatible to dilute gelatins are provided in Table 1
- How long can gelatin be stored?
Gelatin solutions can be stored indefinitely at 2 - 8 °C. Gelatin-coated plates can be stored at 2 - 8 °C for up to 4 weeks provided they are sealed well to prevent contamination and/or over-drying up of gelatin. Do not use the product if discoloration or cracks appear on the surface of the coated material.
- What is the significance of Bloom number for gelatins?
Bloom number was introduced by Oscar T. Bloom and indicates the stiffness of a gelatin film. It was first measured using a Bloom Gelometer by a controlled process to measure the rigidity of a gelatin film. A higher number indicates a stiffer product. The Bloom number/Bloom strength helps in choosing the right gelatin product for your applications.
For additional questions contact our technical services here.