Merck
  • The Dictyostelium type V myosin MyoJ is responsible for the cortical association and motility of contractile vacuole membranes.
The Dictyostelium type V myosin MyoJ is responsible for the cortical association and motility of contractile vacuole membranes.
The Journal of cell biology (2009-08-19)
Goeh Jung, Margaret A Titus, John A Hammer
ABSTRACT
The contractile vacuole (CV) complex in Dictyostelium is a tubulovesicular osmoregulatory organelle that exhibits extensive motility along the actin-rich cortex, providing a useful model for investigating myosin-dependent membrane transport. Here, we show that the type V myosin myoJ localizes to CV membranes and is required for efficient osmoregulation, the normal accumulation of CV membranes in the cortex, and the conversion of collapsed bladder membranes into outwardly radiating cortical CV tubules. Complementation of myoJ-null cells with a version of myoJ containing a shorter lever arm causes these radiating tubules to move at a slower speed, confirming myoJ's role in translocating CV membranes along the cortex. MyoJ-null cells also exhibit a dramatic concentration of CV membranes around the microtubule-organizing center. Consistently, we demonstrate that CV membranes also move bi-directionally on microtubules between the cortex and the centrosome. Therefore, myoJ cooperates with plus and minus end-directed microtubule motors to drive the normal distribution and dynamics of the CV complex in Dictyostelium.